Order Entry
Italy
ContactUsLinkComponent
2-Amino-1,3-propanediol 98%
2-Amino-1,3-propanediol 98%
Certificates
undefined
2-Amino-1,3-propanediol 98%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to off-white Adhering crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    52 to 56 °C
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together