Indocyanine green ≥95%
Supplier: AAT BIOQUEST
Certificates
About this item
Synonyms: IR-125, 4,5-benzoindotricarbocyanine, IC Green, Cardiogreen, Sodium 4-(2-(7-(1,1-dimethyl-3-(4-sulphonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulphonate, Foxgreen
CAS Number:3599-32-4
:
Formula:C₄₃H₄₇N₂NaO₆S₂
MDL Number:MFCD00013078
Molecular Weight:774.99 g/mol
ADR:5 II
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together