Order Entry
Italy
ContactUsLinkComponent
Adenovirus IgM (Human) ELISA kit
Adenovirus IgM (Human) ELISA kit
Catalog # ABNOKA6715.1KIT
Supplier:  Abnova
CAS Number:  
Adenovirus IgM (Human) ELISA kit
Catalog # ABNOKA6715.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Host:
  • Primary antibody reactivity:
    Human
  • Target protein:
    IgM
  • Environmentally Preferable:
  • Assay Principle:
    Qualitative

Specifications

About this item

Adenovirus IgM (Human) ELISA Kit is a qualitative determination of IgM class antibodies against Adenovirus in human serum or plasma.