Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human IL2 & IL15 FUSION (from E. coli)
Recombinant Human IL2 & IL15 FUSION (from E. coli)
Recombinant Human IL2 & IL15 FUSION (from E. coli)
ABNOP7812.20UG
:
Abnova
:
Recombinant Human IL2 & IL15 FUSION (from E. coli)
ABNOP7812.20UG
:
Abnova
:
Pk:
20 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Environmentally Preferable:
Protein/peptide name:
IL2 & IL15 FUSION