Horizontal Electrophoresis Apparatus, M6 and M12
Supplier: EDVOTEK
Certificates
About this item
Capable of running one or two groups of student samples at the same time and producing excellent results in 30 to 40 minutes.
- Seamless, injection-moulded casing
- Safety interlock cover
- Corrosion-resistant platinum electrodes
- Safety insulated electrical leads
- Adjustable leveling feet
Frequently Bought Together