Anti-C2GNT3 Rabbit Polyclonal Antibody (Cy3®)
BOSSBS-15145R-CY3
: Bioss
- Antibody type:Primary
- Antigen name:C2GNT3
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:301-400/453
- Form:Liquid
- Gene ID:51301
- Antigen synonyms:GCNT4|Core2 GlcNAc transferase 3|Core 2 (beta-1,6-N-acetylglucosaminyltransferase)|Glucosaminyl (N-acetyl) transferase 4|Core 2 branching enzyme 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:53 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C2GNT3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Glycosyltransferase that mediates core 2 O-glycan branching, an important step in mucin-type biosynthesis. Does not have core 4 O-glycan or I-branching enzyme activity.
Type: Primary
Antigen: C2GNT3
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 301-400/453
Host: Rabbit
Isotype: IgG
Reactivity: Human