Specifications
- Antibody type:Primary
- Antigen name:Inosine triphosphate pyrophosphatase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:1-100/194
- Form:Liquid
- Gene ID:3704
- Antigen synonyms:My049|My049 protein|OTTHUMP00000030094|Nucleoside triphosphate pyrophosphatase|Itpa|Inosine triphosphatase|C20orf37|dJ794I6.3|NTPase|HLC14-06-P|Inosine triphosphate pyrophosphatase|ITPase|Non canonical purine NTP pyrophosphatase|OTTHUMP00000160459|OK/SW-cl.9|Inosine triphosphatase nucleoside triphosphate pyrophosphatase|nucleoside triphosphate diphosphatase|inosine triphosphatase-A|Putative oncogene protein hlc14-06-p|Non standard purine NTP pyrophosphatase|Inosine triphosphate pyrophosphohydrolase|ITPA_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:21 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Inosine triphosphate pyrophosphatase
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
ITPase (inosine triphosphate pyrophosphatase) is also known as putative oncogene protein hlc14-06-p or ITPA (inosine triphosphatase (nucleoside triphosphate pyrophosphatase)) and is a 194 amino acid protein. ITPase is abundantly expressed in heart, liver, sex glands, thyroid and adrenal gland, and is localized to the cytoplasm in the cell. ITPase catalyzes the pyrophosphohydrolysis of both ITP (inosine triphosphate) and dITP (deoxyinosine triphosphate) to IMP (inosine monophosphate) and diphosphate. IMP can be used as a substrate for purine nucleotide pathways. IMP can be phosphorylated to ITP, and ITPase can regulate the concentration of ITP in the cell by converting ITP back to IMP. Defects in ITPase result in ITPase deficiency which is thought to be inherited and is characterized by an over-accumulation of ITP in erythocytes, leukocytes and fibroblasts.
Type: Primary
Antigen: Inosine triphosphate pyrophosphatase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/194
Host: Rabbit
Isotype: IgG
Reactivity: