Specifications
- Antibody type:Primary
- Antigen name:GCNT3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:351-438/438
- Form:Liquid
- Gene ID:9245
- Antigen synonyms:Core 2 beta 1 6 N acetylglucosaminyltransferase II|hC2GnT-M|C2GnT mucin type|2010013H22Rik|2210401J11Rik|3-galactosyl-O-glycosyl-glycoprotein beta-1|6-N-acetylglucosaminyltransferase|Core 2/core 4 beta 1 6 N acetylglucosaminyltransferase|EC 2.4.1.102|C2GnT-M|C2GNTM|Core 2/core 4 beta-1|Beta-1|C2/4GnT|GCNT3_HUMAN|Glucosaminyl N acetyl transferase 3|GCNT3|hC2GnT M|C24GNT|Glucosaminyl N acetyl transferase 3 mucin type|beta1 6 N acetylglucosaminyltransferase|C2GnT2|Mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase OTTHUMP00000163601|6-N-acetylglucosaminyltransferase 3|EC 2.4.1.150|dI/C2/C4GnT|2210021I22Rik|Beta1 6 N acetylglucosaminyltransferase 3|GNTM|C2GnT-mucin type|C2GnT M|GnT M|Beta 1 3 galactosyl O glycosyl glycoprotein beta 1 6 N acetylglucosaminyltransferase 3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:51 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GCNT3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
2010013H22Rik; 2210021I22Rik; 2210401J11Rik; 3-galactosyl-O-glycosyl-glycoprotein beta-1; 6-N-acetylglucosaminyltransferase 3; 6-N-acetylglucosaminyltransferase; Beta 1 3 galactosyl O glycosyl glycoprotein beta 1 6 N acetylglucosaminyltransferase 3; Beta-1; Beta1 6 N acetylglucosaminyltransferase 3; beta1 6 N acetylglucosaminyltransferase; C2/4GnT; C24GNT; C2GnT M; C2GnT mucin type; C2GnT-M; C2GnT-mucin type; C2GnT2; C2GNTM; Core 2 beta 1 6 N acetylglucosaminyltransferase II; Core 2/core 4 beta 1 6 N acetylglucosaminyltransferase; Core 2/core 4 beta-1; dI/C2/C4GnT; EC 2.4.1.102; EC 2.4.1.150; GCNT3; GCNT3_HUMAN; Glucosaminyl (N acetyl) transferase 3; Glucosaminyl (N acetyl) transferase 3 mucin type; GnT M; GNTM; hC2GnT M; hC2GnT-M; Mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase. OTTHUMP00000163601.
Type: Primary
Antigen: GCNT3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 351-438/438
Host: Rabbit
Isotype: IgG
Reactivity: Mouse