Order Entry
Italy
ContactUsLinkComponent
 
Anti-FOXRED1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-13209R-A680
 :  Bioss
Anti-FOXRED1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-13209R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    FOXRED1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    251-350/486
  • Form:
    Liquid
  • Gene ID:
    55572
  • Antigen synonyms:
    FOXRED1|FAD-dependent oxidoreductase domain-containing protein 1|FXRD1_HUMAN|FAD dependent oxidoreductase domain containing protein 1|FOXRED 1|FP634|FAD dependent oxidoreductase domain containing 1|H17
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    54 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human FOXRED1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

FOXRED1 is a 486 amino acid single-pass membrane protein. Utilizing FAD as a cofactor, FOXRED1 may act as a chaperone protein essential for the function of mitochondrial complex I. Mutations to FOXRED1 may result in mitochondrial complex I deficiency (MT-C1D), which results in a wide range of clinical maladies from lethal neonatal disease to adult onset neurodegenerative disorders. Common phenotypes of MT-C1D include cardiomyopathy, liver disease, Leigh syndrome, Leber hereditary optic neuropathy, and some forms of Parkinson disease. FOXRED1 exists as three alternatively spliced isoforms and is encoded by a gene mapping to human chromosome 11q24.2. With approximately 135 million base pairs and 1,400 genes, chromosome 11 makes up around 4% of human genomic DNA and is considered a gene and disease association dense chromosome.

Type: Primary
Antigen: FOXRED1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-350/486
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat