Anti-Proteasome 20S alpha 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-9355R-A750
: Bioss
- Antibody type:Primary
- Antigen name:Proteasome 20S alpha 6
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:1-100/246
- Form:Liquid
- Gene ID:5687
- Antigen synonyms:Proteasome prosome macropain subunit alpha type 6|Prosomal P27K protein|PROS 27|PROS27|p27K|27 kDa prosomal protein|Macropain subunit iota|Macropain iota chain|Proteasome subunit alpha type 6|PSA6_HUMAN|PSMA6|HC2|Proteasome iota chain|Multicatalytic endopeptidase complex iota chain|Proteasome subunit alpha type 1|IOTA|Proteasome subunit iota
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Proteasome 20S alpha 6
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
The gene encodes a 246-amino acid polypeptide containing an RNA binding motif, a putative nuclear localisation signal, and phosphorylation sites. The alpha subunits comprises the outer rings of the proteasome. Some alpha subunits contain a functional nuclear localisation signal; proteasomes are found in both the nuclear and cytoplasmic compartments of the cell. Alpha subunits may constitute a physical barrier that limits access of cytosolic proteins into the inner proteolytic chamber.
Type: Primary
Antigen: Proteasome 20S alpha 6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/246
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat