Anti-Defensin alpha 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7129R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Defensin alpha 6
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Environmentally Preferable:
- Epitope:31-100/100
- Form:Liquid
- Gene ID:1671
- Antigen synonyms:HD-6|Defensin, alpha 6|Defensin-6|DEF6|DEFA6
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:11 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Defensin alpha 6
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Has very low antimicrobial activity against Gram-negative and Gram-positive bacteria. May protect cells against infection with HIV-1.
Type: Primary
Antigen: Defensin alpha 6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-100/100
Host: Rabbit
Isotype: IgG
Reactivity: Rat