Anti-DDIT4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-4240R-A750
: Bioss
- Antibody type:Primary
- Antigen name:DDIT4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Environmentally Preferable:
- Epitope:165-232/232
- Form:Liquid
- Gene ID:54541
- Antigen synonyms:DDIT4|REDD1|REDD 1|Protein regulated in development and DNA damage response 1|Dig2|DKFZp564O2071|DDIT4_HUMAN|H-1 responsive protein RTP801|FLJ20500|RTP801|REDD-1|DNA damage-inducible transcript 4 protein|DNA damage inducible transcript 4|DNA damage inducible transcript 4 protein|H 1 responsive RTP801|H 1 responsive protein RTP801|RP11 442H21.1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:25 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human DDIT4
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Inhibits cell growth by regulating the TOR signaling pathway upstream of the TSC1-TSC2 complex and downstream of AKT1. Promotes neuronal cell death.
Type: Primary
Antigen: DDIT4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 165-232/232
Host: Rabbit
Isotype: IgG
Reactivity: Rat