Anti-Antithrombin 3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-1636R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Antithrombin 3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:141-211/464
- Form:Liquid
- Gene ID:462
- Antigen synonyms:AT3D|Antithrombin-III|AT3|Serpin C1|SERPINC1|ATIII|THPH7|PRO0309
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:51 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Antithrombin III
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Most important serine protease inhibitor in plasma that regulates the blood coagulation cascade. AT-III inhibits thrombin, matriptase-3/TMPRSS7, as well as factors IXa, Xa and XIa. Its inhibitory activity is greatly enhanced in the presence of heparin.
Type: Primary
Antigen: Antithrombin 3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 141-211/464
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse