Anti-SLC33A1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-0699R-A680
: Bioss
- Antibody type:Primary
- Antigen name:SLC33A1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:481-549/549
- Form:Liquid
- Gene ID:9197
- Antigen synonyms:AT1|CCHLND|AT-1|SPG42|Acetyl-CoA transporter 1|SLC33A1|Solute carrier family 33 member 1|ACATN|Acetyl-coenzyme A transporter 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:61 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SLC33A1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Probable acetyl-CoA transporter necessary for O-acetylation of gangliosides (PubMed:9096318). Negatively regulates BMP signaling (PubMed:25402622).
Type: Primary
Antigen: SLC33A1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 481-549/549
Host: Rabbit
Isotype: IgG
Reactivity: Mouse