Anti-TRA1 Variant Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-0149R-A680
: Bioss
- Antibody type:Primary
- Antigen name:TRA1 variant
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:341-440/803
- Form:Liquid
- Antigen synonyms:TRA1.|tumor rejection antigen (gp96) 1 variant
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:63 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TRA1 variant
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
TRA1 variant.
Type: Primary
Antigen: TRA1 variant
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 341-440/803
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat