Anti-eEF1A1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-2503R-A680
: Bioss
- Antibody type:Primary
- Antigen name:eEF1A1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:131-230/462
- Form:Liquid
- Gene ID:1915
- Antigen synonyms:Leukocyte receptor cluster member 7|Eukaryotic elongation factor 1 A-1|EEF-1|EF1A|EE1A1|GRAF-1EF|CCS-3|EF-Tu|LENG7|PTI1|eEF1A-1|HNGC:16303|EEF1A|EEF1A1|EF-1-alpha-1|Elongation factor Tu|CCS3|Elongation factor 1-alpha 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:50 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human eEF1A1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
This protein promotes the GTP-dependent binding of aminoacyl-tRNA to the A-site of ribosomes during protein biosynthesis. With PARP1 and TXK, forms a complex that acts as a T helper 1 (Th1) cell-specific transcription factor and binds the promoter of IFN-gamma to directly regulate its transcription, and is thus involved importantly in Th1 cytokine production.
Type: Primary
Antigen: eEF1A1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 131-230/462
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat