Order Entry
Italy
ContactUsLinkComponent
 
Anti-C4 binding protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-15184R-A750
 :  Bioss
Anti-C4 binding protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-15184R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    C4 binding protein
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    1-100/252
  • Form:
    Liquid
  • Antigen synonyms:
    PRP|OTTHUMP00000034375|C4b-binding protein beta chain|OTTHUMP00000034288|OTTHUMP00000034289|OTTHUMP0000003474|Complement component 4-binding protein beta|Complement component 4 binding protein beta chain|C4BP|Complement component 4 binding protein beta|C4b binding protein beta chain|C4BPB_HUMAN|Proline rich protein|C4BPB
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    26 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human C4 binding protein
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

C4BPB is a member of a superfamily of proteins composed predominantly of tandemly arrayed short consensus repeats of approximately 60 amino acids. A single, unique beta-chain encoded by this gene assembles with seven identical alpha-chains into the predominant isoform of C4b-binding protein, a multimeric protein that controls activation of the complement cascade through the classical pathway. C4b-binding protein has a regulatory role in the coagulation system also, mediated through the beta-chain binding of protein S, a vitamin K-dependent protein that serves as a cofactor of activated protein C. The genes encoding both alpha and beta chains are located adjacent to each other on human chromosome 1 in the regulator of complement activation gene cluster. Alternative splicing gives rise to multiple transcript variants.

Type: Primary
Antigen: C4 binding protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/252
Host: Rabbit
Isotype: IgG
Reactivity: Human