Specifications
- Antibody type:Primary
- Antigen name:FBP17
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pufferfish,Xenopus laevis
- Western blot:Yes
- Environmentally Preferable:
- Epitope:331-430/617
- Form:Liquid
- Gene ID:23048
- Antigen synonyms:Formin-binding protein 17|FNBP1_HUMAN|KIAA0554|MGC126804|hFBP17|FBP1|Formin binding protein 17|Formin-binding protein 1|FNBP1|Formin binding protein 1|FBP17
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:71 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human FBP17
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
FNBP1 is a 617 amino acid protein that localizes to a variety of locations within the cell, including the cytoplasm, cytoskeleton, lysosome and the cell cortex, and contains one FCH domain, one REM repeat and one SH3 domain. Expressed at high levels in respiratory, reproductive and urinary systems, as well as in brown adipose tissue and epithelial cells of the gastrointestinal tract, FNBP1 interacts with Rho 7 and links the Actin cytoskeleton with Rho 7 signaling, playing a crucial role in membrane tubulation and cytoskeletal reorganization during endocytosis. Additionally, FNBP1, which exists as four alternatively spliced isoforms, enhances Actin polymerization and promotes membrane invagination and the formation of tubules. Chromosomal aberrations in the FNBP1 gene are associated with acute leukemias, suggesting a role for defective FNBP1 in carcinogenesis.
Type: Primary
Antigen: FBP17
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 331-430/617
Host: Rabbit
Isotype: IgG
Reactivity: Xenopus laevis; Pufferfish