Anti-Nrf2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-32822
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Respiratory Factor 2
- Antigen symbol:Nrf2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:4780
- Antigen synonyms:NFE2-related factor 2|HEBP1|Nrf2 rat|Nrf2 polyclonal|Nrf2 immunofluorescence|nuclear factor erythroid-derived 2-like 2|nuclear factor erythroid 2-related factor 2|NF-E2-related factor 2|NRF2 erythroid derived 2|Nrf2 immunohistochemistry|Nrf2 western blot|nuclear factor (erythroid-derived 2)-like 2|like 2|Nrf2 ihc
- Storage buffer:PBS (pH 7,0) and 20% Glycerol
- Storage temperature:Aliquot and store at ‒20 °C or ‒80 °C. Avoid freeze-thaw cycles.
- Concentration:0,51 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human NRF2. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Pk:100 µl
Specifications
About this item
The Nrf2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Nrf2. This antibody reacts with human, mouse, rat. The Nrf2 Antibody has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100 to 1:1000
Type: Primary
Antigen: Nrf2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat