Anti-ARK5/NUAK1 Goat Polyclonal Antibody
NOVUNBP1-36937
- Antibody type:Primary
- Antigen name:NUAK Family SNF1-like kinase 1
- Antigen symbol:ARK5/NUAK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal region
- Gene ID:9891
- Antigen synonyms:ARK5AMP-activated protein kinase family member 5|NUAK family|KIAA0537NUAK family SNF1-like kinase 1|EC 2.7.11.1|NuaK1|SNF1-like kinase|EC 2.7.11|1|AMPK-related protein kinase 5
- Storage buffer:0,5 mg/ml Tris (pH 7,3) and 0,5% BSA
- Storage temperature:Store at ‒20 °C. Avoid freeze-thaw cycles.
- Concentration:0,5 mg/ml
- Immunogen:Peptide with sequence C-QNRPRPQYLKRYRNR corresponding to internal region according to NP_055655.1.
- Purification:Immunogen affinity purified
- Pk:100 µl
The ARK5 / NUAK1 Antibody from Novus Biologicals is a goat polyclonal antibody to ARK5 / NUAK1. This antibody reacts with human. The ARK5 / NUAK1 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Peptide
Type: Primary
Antigen: ARK5/NUAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal region
Host: Goat
Isotype: IgG
Reactivity: Human