Fetal Bovine Serum, Gold, Heat Inactivated, MP Biomedicals
Catalog # ICNA092916849
Supplier: MP Biomedicals
Specifications
- Sterility:Sterile
- Description:Fetal Bovine Serum, Gold, Heat Inactivated, CELLect™, 100 ml
- Environmentally Preferable:
- Endotoxin:0,7 EU/ml
- Storage temperature:Store at –20 °C
- Application:Source of growth promoting and survival enhancing factor.
- Concentration:Protein: 3,1 - 4,4%
- Pk:100 mL
Specifications
About this item
Fetal bovine serum has been the most widely used growth supplement for cell culture media because of its high content of embryonic growth-promoting factors.
- Supplies components that satisfy specific metabolic requirements for the culture of cells in vitro
- Suitable for cell culture and insect cell culture
- Source of growth promoting and survival enhancing factor
CELLect™ FBS, Gold, Heat Inactivated is used in a broad range of cell culture applications.
Serum is an extremely complex mixture of many small and large biomolecules with different, physiologically balanced growth-promoting and growth-inhibiting activities. Some of the major functions of serum are to provide growth factors, hormones, attachment and spreading factors, binding proteins, lipids, and minerals.
FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture.