Multiwell cell culture plates, Falcon®
Supplier: Corning
Certificates
About this item
Standard tissue culture (TC) and Corning® Primaria™ surface treatments is available. These plates are provided with various colours to suit various detection methods.
- Crystal-grade virgin polystyrene
- Nonpyrogenic
- Sterilised by gamma irradiation
- Growth area and volume are nominal
Individual and ready-stack (RS) trays are PET (Code 1) and recyclable.
Packaging: Convenient, peel-open medical-style packaging for individual product presentation.
Frequently Bought Together