Water, sterile distilled
Supplier: G-Biosciences
Certificates
About this item
Synonyms: Hydrogen oxide
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
Boiling Pt:100
Molecular Weight:18.02 g/mol
Melting Pt:0
Merck Index:13,10098
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together