Lithium tetrafluoroborate, anhydrous 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14283-07-9
:
Formula:BF₄Li
MDL Number:MFCD00011087
Molecular Weight:93.74 g/mol
Melting Pt:309…311 °C
Storage Temperature:Ambient
EINECS:238-178-9
UN:3260
ADR:8 II
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together