2,6-Difluoro-4-methoxyphenylboronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:406482-20-0
:
Formula:C₇H₇BF₂O₃
MDL Number:MFCD05664299
Molecular Weight:187.94 g/mol
Melting Pt:125…130 °C
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together