5-Formylpyridine-3-boronic acid pinacol ester
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinaldehyde
CAS Number:848093-29-8
:
Formula:C₁₂H₁₆BNO₃
MDL Number:MFCD04038759
Molecular Weight:233.074 g/mol
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together