1H-Pyrazole-5-carbaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1H-Pyrazol-5-carbaldehyde, 5-Formyl-1H-pyrazole
CAS Number:948552-36-1
:3920-50-1
Formula:C₄H₄N₂O
MDL Number:MFCD06657574
Molecular Weight:96.087 g/mol
:GHS05
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together