(R)-(-)-Prolinol 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: D-Prolinol, (2R)-pyrrolidin-2-ylmethanol, D(-)-Prolinol
CAS Number:68832-13-3
:
Formula:C₅H₁₁NO
MDL Number:MFCD00064321
Boiling Pt:74…76 °C/2mmHg
Density:1.025 g/cm³ (25 °C)
Molecular Weight:101.1 g/mol
Flash Pt:86 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together