D-(+)-Sucrose Pure BP, Ph. Eur., JP, USP, pharma grade
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
CAS Number:57-50-1
:
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006626
Molecular Weight:342.3 g/mol
Melting Pt:186 °C
Merck Index:13,08966
Storage Temperature:Ambient
EINECS:200-334-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together