3-(Cyclopropyl)thiophene-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:29488-33-3
:
Formula:C₈H₈O₂S
MDL Number:MFCD13195793
Molecular Weight:168.213 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together