Order Entry
Italy
ContactUsLinkComponent
Rhodamine B base 97% (dye content), Sigma-Aldrich®
Rhodamine B base 97% (dye content), Sigma-Aldrich®
Certificates
Rhodamine B base 97% (dye content), Sigma-Aldrich®
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance (Form):
    Powder or Chunks
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength, C=0.0025 g/L in Methanol:
    542 - 546 nm
  • Extinction Coefficient:
    ≥103000
  • Carbon:
    73.3 - 78.6%
  • Nitrogen:
    6.1 - 6.6%
  • Purity (HPLC):
    ≥96.5%
  • Solubility (Turbidity), c = 1 mg/ml; Ethanol:
    Clear to Slightly Hazy
  • Solubility (Colour):
    Orange and Red to Dark Red and Orange-Red and Red-Orange

Specification Test Results

Related Information

Frequently Bought Together