Crystal violet 1% in aqueous solution, Sigma-Aldrich®
: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
:548-62-9
:
:C₂₅H₃₀ClN₃
:MFCD00011750
:407.99 g/mol
:13,04406
:M-Clarity™ quality level = MQ200
:GHS08
:GHS07
:Room Temperature
- Environmentally Preferable:
- Appearance (Form):Powder
- Solubility (Colour):Faint Red to Very Dark Red to Very Dark Purple
- Solubility (Form)
10 mg/mL , 1M NH4OH in MeOH:Solution - 1H NMR Spectrum:Conforms to Structure
- Solvent Content (by GC):≤ 4 %
- Nitrogen:≥7.5 %
- Dye Content:≥80 %
- Labeling Efficiency:≥50 %
Frequently Bought Together