Strontium perchlorate trihydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15650-09-6
:
Formula:Cl₂O₈Sr·3H₂O
MDL Number:MFCD09039117
Molecular Weight:340.57 g/mol
Storage Temperature:Ambient
EINECS:236-614-2
UN:1508
ADR:5.1 II
:GHS03
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together