4-Fluoro-2-iodoaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:61272-76-2
:
Formula:C₆H₅FIN
MDL Number:MFCD00042584
Molecular Weight:237.02 g/mol
UN:2810
ADR:6.1 III
:GHS06
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together