Tris(4-biphenylyl)amine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
: TCI
: Tri([1,1'-biphenyl]-4-yl)amine
:6543-20-0
:
:C₃₆H₂₇N
:MFCD30470498
:473.62 g/mol
- Environmentally Preferable:
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Frequently Bought Together