Thymidine-5'-monophosphate disodium salt hydrate ≥98.0% (by HPLC)
Catalog # TCIAT0845-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 93.0 %
- Specific rotation [a]20/D:-6.0 to -9.0 deg (C=0.4, H2O)
- abs-ratio (a/b):0.62 to 0.66 (E250/E260)
Specification Test Results
About this item
Synonyms: dTMP 2Na, 5'-Thymidylic Acid Disodium Salt
:
:366.17 g/mol
Storage Temperature:Refrigerator
EINECS:251-518-0
CAS Number:33430-62-5
Formula:C₁₀H₁₅N₂O₈P
MDL Number:MFCD00023797