SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate) ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
Synonyms: 2,5-Dioxopyrrolidin-1-yl 3-(pyridin-2-yldisulphanyl)propanoate, 3-(2-Pyridyldithio)propionic Acid N-Succinimidyl Ester, SPDP, 3-(2-Pyridyldithio)-Propionic Acid N-Hydroxysuccinimide Ester
CAS Number:68181-17-9
:
Formula:C₁₂H₁₂N₂O₄S₂
MDL Number:MFCD00009636
Molecular Weight:312.36 g/mol
Storage Temperature:Freezer
EINECS:269-034-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:87.0 to 91.0 deg-C
Specification Test Results
Frequently Bought Together