Penicillin G potassium salt
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: 6-(2-Phenylacetamido)penicillanic acid potassium salt, Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin G potassium, Potassium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Benzylpenicillin potassium salt
CAS Number:113-98-4
:
Formula:C₁₆H₁₇KN₂O₄S
MDL Number:MFCD00036193
Density:1.359 g/cm³ (20 °C)
Molecular Weight:372.49 g/mol
Melting Pt:214…217 °C
Merck Index:12,07225
EINECS:204-038-0
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together