4-Sulphophthalic acid ∼50% in water (contains 3-Sulfophthalic Acid)
Supplier: TCI
Certificates
About this item
CAS Number:89-08-7
:
Formula:C₈H₆O₇S
MDL Number:MFCD00007494
Molecular Weight:246.19 g/mol
Storage Temperature:Refrigerator
EINECS:201-881-6
UN:3265
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (HPLC):min. 80.0 area%
- Concentration (Neutralization titration):50.0 to 60.0 w/w%
- Water:40.0 to 50.0 %
- Ignition residue (Sulfate):max. 0.1 %
Specification Test Results
Frequently Bought Together