Order Entry
Italy
ContactUsLinkComponent
2-(1H-1,2,4-Triazol-3-yl)pyridine ≥98.0% (by GC, titration analysis)
2-(1H-1,2,4-Triazol-3-yl)pyridine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
2-(1H-1,2,4-Triazol-3-yl)pyridine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    160.0 to 164.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together