Benzophenone-4-maleimide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 1-(4-Benzoylphenyl)-1H-pyrrole-2,5-dione, 4-(N-Maleimido)benzophenone, Benzophenone-4-maleimide
CAS Number:92944-71-3
:
Formula:C₁₇H₁₁NO₃
MDL Number:MFCD00079465
Molecular Weight:277.2700 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:158.0 to 162.0 deg-C
Specification Test Results
Frequently Bought Together