(±)-3-Methyl-2-pentanol mixture of diasteroisomers ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 3-Methyl-2-pentanol
CAS Number:565-60-6
:
Formula:C₆H₁₄O
MDL Number:MFCD00004528
Boiling Pt:134
Molecular Weight:102.18 g/mol
Melting Pt:-85
Flash Pt:40°C
EINECS:209-281-6
UN:1987
ADR:3 III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of diastereoisomers)
Specification Test Results
Frequently Bought Together