Order Entry
Italy
ContactUsLinkComponent
4-(4-Heptylphenyl)benzoic acid ≥95.0% (by GC)
4-(4-Heptylphenyl)benzoic acid ≥95.0% (by GC)
Catalog # TCIAH0709-5G
Supplier:  TCI
CAS Number:  
4-(4-Heptylphenyl)benzoic acid ≥95.0% (by GC)
Catalog # TCIAH0709-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Solubility in hot EtOH:
    almost transparency

Specification Test Results

About this item

Synonyms: 4-Heptylbiphenyl-4'-carboxylic Acid
 :
 :296.41 g/mol
 :GHS07
CAS Number:58573-94-7
Formula:C₂₀H₂₄O₂
MDL Number:MFCD00143231