Z-Aib-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: N-Benzyloxycarbonyl-alpha-methylalanine, 2-(Benzyloxycarbonylamino)isobutyric acid, N-Benzyloxycarbonyl-2-methylalanine, Z-Aib-OH, 2-(Cbz-amino)isobutyric acid, 2-(Carbobenzoxyamino)isobutyric acid, N-Cbz-alpha-methylalanine
CAS Number:15030-72-5
:
Formula:C₁₂H₁₅NO₄
MDL Number:MFCD00004190
Molecular Weight:237.255 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:77.0 to 81.0 deg-C
Specification Test Results
Frequently Bought Together