Alimemazine Tartrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: Trimeprazine Tartrate, 10-[3-(Dimethylamino)-2-methylpropyl]phenothiazine Tartrate, Alimemazine Hemitartrate
CAS Number:4330-99-8
:
Formula:C₃₆H₄₄N₄S₂·C₄H₆O₆
MDL Number:MFCD00242594
Molecular Weight:746.98 g/mol
ADR:6.1 III
Merck Index:14,09704
EINECS:224-368-9
UN:2811
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:159.0 to 163.0 deg-C
Specification Test Results
Frequently Bought Together