Dibutyl adipate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Adipic Acid Dibutyl Ester
CAS Number:105-99-7
:
Formula:C₁₄H₂₆O₄
MDL Number:MFCD00009451
Boiling Pt:305°C
Molecular Weight:258.36 g/mol
Melting Pt:–32°C
Flash Pt:166°C
EINECS:203-350-4
UN:3082
ADR:9 III
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
- Acid Value:max. 0.5
Specification Test Results
Frequently Bought Together