Calcium lactate pentahydrate, soluble, EMPROVE® ESSENTIAL E327, FCC, BP, Ph. Eur., JP, USP, extra pure suitable for use as excipient, SAFC®
Catalog # 1.02102.9029
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
- Environmentally Preferable:
Specifications
About this item
Synonyms: L(+)-Lactic acid calcium salt pentahydrate, Calcium (S)-(+)-2-hydroxypropionate pentahydrate, (S)-(+)-2-Hydroxypropionic acid calcium salt pentahydrate, (S)-(+)-Lactic acid calcium salt pentahydrate, (S)-(+)-2-Hydroxypropanoic acid calcium salt pentahydrate, Calcium L(+)-2-hydroxypropionate pentahydrate
:
:308.3 g/mol
Melting Pt:240
Storage Temperature:Ambient
EINECS:248-953-3
CAS Number:5743-47-5
Formula:C₆H₁₀CaO₆·5H₂O
MDL Number:MFCD00150344