Sodium hypochlorite (6 - 14% Cl₂) in aqueous solution, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7681-52-9
:
Formula:ClNaO
MDL Number:MFCD00011120
Density:1.22…1.25 g/cm³ (20 °C)
Molecular Weight:74.44 g/mol
Melting Pt:–30…–20 °C
ADR:8 II
Merck Index:13,08702
:M-Clarity™ quality level = MQ200
EINECS:231-668-3
UN:1791
:GHS05
:GHS09
Storage Temperature:Refrigerator
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together