Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Inorganic disinfection by-product kit, (CRM), Supelco®
Inorganic disinfection by-product kit, (CRM), Supelco®
Inorganic disinfection by-product kit, (CRM), Supelco®
Catalog # PE1306-1KT
Supplier:
Merck
CAS Number:
Expand image
Inorganic disinfection by-product kit, (CRM), Supelco®
Catalog # PE1306-1KT
Supplier:
Merck
CAS Number:
Specifications
Pk:
1 KIT
Description:
INORGANIC DISINFECTION BY-PRODUCTS KIT -
Environmentally Preferable:
Specifications
About this item
Product Details
Product Details