Exonuclease I
Supplier: Thermo Fisher Scientific
Certificates
About this item
Exonuclease I (ExoI) degrades single-stranded DNA in a 3' to 5'direction. It releases deoxyribonucleoside 5'-monophosphates in a stepwise manner and leaves 5'-terminal dinucleotides intact.
Used for primer removal from PCR mixtures, prior to PCR product sequencing, for one-tube 'megaprimer' PCR mutagenesis, removal of single-stranded DNA containing a 3'-hydroxyl terminus from nucleic acid mixtures, assay for the presence of single-stranded DNA with a 3'-hydroxyl terminus.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together